accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86060-93-7

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 3-[4-(tert-butoxy)phenyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate
Molecular Weight (g/mol): 625.59
Molecular Formula: C34H28F5NO5
InChi Key: ADOSTZDWXCEVJP-UHFFFAOYNA-N
SMILES: CC(C)(C)OC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(=O)OC2=C(F)C(F)=C(F)C(F)=C2F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results