accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86061-01-0

IUPAC Name: 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanedioate
Synonyms: 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanedioate
Molecular Weight (g/mol): 577.5
Molecular Formula: C29H24F5NO6
InChi Key: DWYWJUBBXKYAMY-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)