CAS RN 860653-59-4
IUPAC Name:
potassium (fluorosulfonyl)(trifluoromethanesulfonyl)azanide
Synonyms:
potassium (fluorosulfonyl)(trifluoromethanesulfonyl)azanide
Molecular Weight (g/mol):
269.23
Molecular Formula:
CF4KNO4S2
InChi Key:
IMBHGOLFXVHBMC-UHFFFAOYSA-N
SMILES:
[K+].FC(F)(F)S(=O)(=O)[N-]S(F)(=O)=O
1
–
1
of
1
results
1
Potassium (Fluorosulfonyl)(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
CAS: 860653-59-4 Molecular Formula: CF4KNO4S2 Molecular Weight (g/mol): 269.23 MDL Number: MFCD28334505 InChI Key: IMBHGOLFXVHBMC-UHFFFAOYSA-N PubChem CID: 45378297 IUPAC Name: potassium (fluorosulfonyl)(trifluoromethanesulfonyl)azanide SMILES: [K+].FC(F)(F)S(=O)(=O)[N-]S(F)(=O)=O