accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86087-24-3

IUPAC Name: oxolan-3-ol
Synonyms: 3-hydroxytetrahydrofuran
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: XDPCNPCKDGQBAN-UHFFFAOYNA-N
SMILES: OC1CCOC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)