accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CH3CH3CH3CSBOOCH3CH3CH3CH3S

CAS RN 861398-06-3

IUPAC Name: 4,4,5,5-tetramethyl-2-[11-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[11-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 442.2
Molecular Formula: C22H28B2O4S2
InChi Key: NTSDQKBGUDHDGQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=CC3=C(C=C(S3)B3OC(C)(C)C(C)(C)O3)C=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results