accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86180-54-3

IUPAC Name: 3-({2-[(2-cyanoethyl)sulfanyl]ethyl}sulfanyl)propanenitrile
Synonyms: 3-({2-[(2-cyanoethyl)sulfanyl]ethyl}sulfanyl)propanenitrile
Molecular Weight (g/mol): 200.32
Molecular Formula: C8H12N2S2
InChi Key: SJYFOHJCZZEPNC-UHFFFAOYSA-N
SMILES: N#CCCSCCSCCC#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results