accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 862095-77-0

BFFFFNNNFFF

CAS RN 862095-77-0

IUPAC Name: 5-benzyl-2-[4-(trifluoromethyl)phenyl]-2H,5H,6H,7H-4λ⁵-pyrrolo[2,1-c][1,2,4]triazol-4-ylium; tetrafluoroboranuide
Synonyms: 5-benzyl-2-[4-(trifluoromethyl)phenyl]-5H,6H,7H-4λ⁵-pyrrolo[2,1-c][1,2,4]triazol-4-ylium tetrafluoroborate
Molecular Weight (g/mol): 431.16
Molecular Formula: C19H17BF7N3
InChi Key: UDDOGJQUYCEOBY-UHFFFAOYNA-N
SMILES: F[B-](F)(F)F.FC(F)(F)C1=CC=C(C=C1)N1C=[N+]2C(CC3=CC=CC=C3)CCC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results