accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 862121-57-1

IUPAC Name: strontium(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms: strontium(2+) bis(bistriflylimide anion)
Molecular Weight (g/mol): 647.89
Molecular Formula: C4F12N2O8S4Sr
InChi Key: DIMYICIXZTVMOY-UHFFFAOYSA-N
SMILES: [Sr++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results