accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 862129-81-5

IUPAC Name: 2-(3,6-dihydro-2H-thiopyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,6-dihydro-2H-thiopyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 226.14
Molecular Formula: C11H19BO2S
InChi Key: QZVRTORVCKGPPY-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CCSCC1
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results