accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFFFFFFCu

CAS RN 86233-74-1

IUPAC Name: (1Z,5Z)-cycloocta-1,5-diene (3Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one copper
Synonyms: 1,5-cyclooctadiene, (Z,Z)- hexafluoroacetylacetone copper
Molecular Weight (g/mol): 379.79
Molecular Formula: C13H14CuF6O2
InChi Key: WSAPIYREFIKPSH-OJKFPHAYSA-N
SMILES: [Cu].C1C\C=C/CC\C=C/1.O\C(=C/C(=O)C(F)(F)F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results