accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86270-03-3

IUPAC Name: 3-(trifluoromethoxy)benzoyl chloride
Synonyms: 3-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: MGKIOAXLPYJSMU-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC(=C1)C(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results