accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86308-26-1

IUPAC Name: 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 212.14
Molecular Formula: C12H25BO2
InChi Key: LDPMCSWZEGKWLC-UHFFFAOYSA-N
SMILES: CCCCCCB1OC(C)(C)C(C)(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results