CAS RN 86356-73-2
IUPAC Name:
methylidene[(2,3,4,5,6-pentafluorophenyl)methoxy]amine
Synonyms:
methylidene[(2,3,4,5,6-pentafluorophenyl)methoxy]amine
Molecular Weight (g/mol):
225.12
Molecular Formula:
C8H4F5NO
InChi Key:
SRTQFRQWTUMMTC-UHFFFAOYSA-N
SMILES:
FC1=C(F)C(F)=C(CON=C)C(F)=C1F
1
–
1
of
1
results
1
O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime 98.0+%, TCI America™
CAS: 86356-73-2 Molecular Formula: C8H4F5NO Molecular Weight (g/mol): 225.118 MDL Number: MFCD00191477 InChI Key: SRTQFRQWTUMMTC-UHFFFAOYSA-N PubChem CID: 602486 IUPAC Name: N-[(2,3,4,5,6-pentafluorophenyl)methoxy]methanimine SMILES: C=NOCC1=C(C(=C(C(=C1F)F)F)F)F