accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BOH3CH3CH3CCH3ONH2

CAS RN 863578-36-3

IUPAC Name: 5-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 5-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 233.12
Molecular Formula: C13H20BNO2
InChi Key: PEXRISJTWBBZRY-UHFFFAOYSA-N
SMILES: CC1=CC=C(B2OC(C)(C)C(C)(C)O2)C(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results