accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 863753-30-4

IUPAC Name: (2-aminophenyl)boronic acid hydrochloride
Synonyms: 2-aminophenylboronic acid hydrochloride
Molecular Weight (g/mol): 173.4
Molecular Formula: C6H9BClNO2
InChi Key: WPDASZCYRKGSTO-UHFFFAOYSA-N
SMILES: Cl.NC1=CC=CC=C1B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)