CAS RN 86390-77-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ 1-Oleoyl-2-acetyl-sn-glycerol
Cell-permeable diacylglycerol analog that activates Ca2+-dependent protein kinase C.