accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 864068-82-6

IUPAC Name: ({imidazo[1,2-a]pyridin-6-yl}methyl)(methyl)amine
Synonyms: {imidazo[1,2-a]pyridin-6-ylmethyl}(methyl)amine
Molecular Weight (g/mol): 161.21
Molecular Formula: C9H11N3
InChi Key: VTAYGJSBPFKWJX-UHFFFAOYSA-N
SMILES: CNCC1=CN2C=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results