accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 864068-85-9

IUPAC Name: 2,4-diphenyl-1,3-thiazole-5-carbaldehyde
Synonyms: 2,4-diphenyl-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 265.33
Molecular Formula: C16H11NOS
InChi Key: QCAWTHNKWALNJR-UHFFFAOYSA-N
SMILES: O=CC1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results