accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNOH

CAS RN 864068-86-0

IUPAC Name: (2,4-diphenyl-1,3-thiazol-5-yl)methanol
Synonyms: (2,4-diphenyl-1,3-thiazol-5-yl)methanol
Molecular Weight (g/mol): 267.35
Molecular Formula: C16H13NOS
InChi Key: IVIJHUHOSCRMNY-UHFFFAOYSA-N
SMILES: OCC1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results