accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNNOO

CAS RN 864754-17-6

IUPAC Name: 1-[(1,3-dioxolan-2-yl)methyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-(1,3-dioxolan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 280.13
Molecular Formula: C13H21BN2O4
InChi Key: IWEGDQUCWQFKHS-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN(CC2OCCO2)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results