accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 864754-21-2

IUPAC Name: 3-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]methyl}pyridine
Synonyms: 3-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methyl}pyridine
Molecular Weight (g/mol): 285.15
Molecular Formula: C15H20BN3O2
InChi Key: LVLQAPQDEZOFQU-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN(CC2=CN=CC=C2)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results