accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3CH3CH3NNOOH3CCH3H3C

CAS RN 864771-44-8

IUPAC Name: tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
Synonyms: tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole-1-carboxylate
Molecular Weight (g/mol): 344.22
Molecular Formula: C18H25BN2O4
InChi Key: MMXDKXUKSKMROG-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1N=CC2=CC(=CC=C12)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results