accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CS(R)(R)NHO(R)OHOSON(S)HOOOHO(R)ON(R)OHO(Z)

CAS RN 86482-18-0

IUPAC Name: (2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid; (2S,5R,6R)-6-[(2R)-2-carboxy-2-(thiophen-3-yl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Synonyms: ticarcillin clavulanate
Molecular Weight (g/mol): 583.58
Molecular Formula: C23H25N3O11S2
InChi Key: XWMVMWTVLSLJGY-FAJPTIRJSA-N
SMILES: OC\C=C1/O[C@@H]2CC(=O)N2[C@H]1C(O)=O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](C(O)=O)C3=CSC=C3)C(=O)N2[C@H]1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results