accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 865-34-9

IUPAC Name: lithium(1+) methanolate
Synonyms: lithium methoxide
Molecular Weight (g/mol): 37.97
Molecular Formula: CH3LiO
InChi Key: JILPJDVXYVTZDQ-UHFFFAOYSA-N
SMILES: [Li+].C[O-]
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)