accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClIClCl

CAS RN 865-44-1

IUPAC Name: trichloro-λ³-iodane
Synonyms: iodine trichloride
Molecular Weight (g/mol): 233.25
Molecular Formula: Cl3I
InChi Key: PAWIVBWALDNUJP-UHFFFAOYSA-N
SMILES: Cl[I](Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)