accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3GeCH3CH3H3C

CAS RN 865-52-1

IUPAC Name: tetramethylgermane
Synonyms: tetramethyl germane
Molecular Weight (g/mol): 132.77
Molecular Formula: C4H12Ge
InChi Key: ZRLCXMPFXYVHGS-UHFFFAOYSA-N
SMILES: C[Ge](C)(C)C
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)