accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86522-89-6

IUPAC Name: 2,4-dichloro-5-fluorobenzoate
Synonyms: 2,4-dichloro-5-fluorobenzoate
Molecular Weight (g/mol): 207.99
Molecular Formula: C7H2Cl2FO2
InChi Key: KZCWJHUTTSVCRO-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=C(Cl)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results