accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 865869-28-9

IUPAC Name: (4,4-dimethylcyclohex-1-en-1-yl)boronic acid
Synonyms: 4,4-dimethylcyclohex-1-en-1-ylboronic acid
Molecular Weight (g/mol): 154.02
Molecular Formula: C8H15BO2
InChi Key: HWFBGGLUEZIDOE-UHFFFAOYSA-N
SMILES: CC1(C)CCC(=CC1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results