CAS RN 866-55-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
249.75
Molecular Formula:
C4H12Cl2Sn
InChi Key:
RFGFYUBDZRGQMZ-UHFFFAOYSA-N
SMILES:
Cl.Cl.CC[SnH2]CC
1
–
1
of
1
results
1
Diethyltin dichloride
CAS: 866-55-7 Molecular Formula: C4H10Cl2Sn Molecular Weight (g/mol): 247.734 MDL Number: MFCD00013609 InChI Key: YFAXVVMIXZAKSR-UHFFFAOYSA-L Synonym: diethyltin dichloride,diethyldichlorotin,diethyltindichloride,dichlorodiethyltin,diethyldichlorostannane,tin, dichlorodiethyl,stannane, dichlorodiethyl,diethyltin chloride,tin, diethyl-, dichloride,unii-pga900d045 PubChem CID: 91491 IUPAC Name: dichloro(diethyl)stannane SMILES: CC[Sn](CC)(Cl)Cl