accessibility menu, dialog, popup

UserName

Viewing profile as user:
KHOOOHOOOHO

CAS RN 866-83-1

IUPAC Name: potassium 3,4-dicarboxy-3-hydroxybutanoate
Synonyms: potassium 3,4-dicarboxy-3-hydroxybutanoate
Molecular Weight (g/mol): 230.21
Molecular Formula: C6H7KO7
InChi Key: WKZJASQVARUVAW-UHFFFAOYNA-M
SMILES: [K+].OC(=O)CC(O)(CC([O-])=O)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)