accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 866089-28-3

IUPAC Name: 2,2-dimethyl-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 2,2-dimethyl-3,4-dihydro-1,4-benzoxazine
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: WILBVUMFTFLAHJ-UHFFFAOYSA-N
SMILES: CC1(C)CNC2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)