accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNIFFF

CAS RN 866638-72-4

IUPAC Name: 4-iodo-5-(trifluoromethyl)-1H-pyrazole
Synonyms: 4-iodo-3-(trifluoromethyl)-2H-pyrazole
Molecular Weight (g/mol): 261.97
Molecular Formula: C4H2F3IN2
InChi Key: XRUHSOXIZMYOHX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(I)C=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results