accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86672-58-4

IUPAC Name: 6-methyl-2-[(4-methylphenyl)amino]-4H-3,1-benzoxazin-4-one
Synonyms: 6-methyl-2-[(4-methylphenyl)amino]-3,1-benzoxazin-4-one
Molecular Weight (g/mol): 266.3
Molecular Formula: C16H14N2O2
InChi Key: GFWNGVKCDGYFKG-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC2=NC3=CC=C(C)C=C3C(=O)O2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results