accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86770-67-4

IUPAC Name: 2-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}ethan-1-ol
Synonyms: 2-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}ethanol
Molecular Weight (g/mol): 219.24
Molecular Formula: C8H17N3O4
InChi Key: MBQYGQMGPFNSAP-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCN=[N+]=[N-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results