accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86776-52-5

H3COONF

CAS RN 86776-52-5

IUPAC Name: 4-cyano-3-fluorophenyl 4-butylbenzoate
Synonyms: 4-cyano-3-fluorophenyl 4-butylbenzoate
Molecular Weight (g/mol): 297.33
Molecular Formula: C18H16FNO2
InChi Key: IBUUMDHMTARCSQ-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C(=O)OC1=CC(F)=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results