accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNClONHN

CAS RN 868273-06-7

IUPAC Name: 5-chloro-3-ethyl-N-{2-[4-(piperidin-1-yl)phenyl]ethyl}-1H-indole-2-carboxamide
Synonyms: 5-chloro-3-ethyl-N-{2-[4-(piperidin-1-yl)phenyl]ethyl}-1H-indole-2-carboxamide
Molecular Weight (g/mol): 409.96
Molecular Formula: C24H28ClN3O
InChi Key: AHFZDNYNXFMRFQ-UHFFFAOYSA-N
SMILES: CCC1=C(NC2=CC=C(Cl)C=C12)C(=O)NCCC1=CC=C(C=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results