CAS RN 86847-71-4
IUPAC Name:
N-(3-formylpyridin-4-yl)-2,2-dimethylpropanamide
Synonyms:
N-(3-formylpyridin-4-yl)-2,2-dimethylpropanamide
Molecular Weight (g/mol):
206.25
Molecular Formula:
C11H14N2O2
InChi Key:
ICMXCEJBHWHTBH-UHFFFAOYSA-N
SMILES:
CC(C)(C)C(=O)NC1=CC=NC=C1C=O
1
–
1
of
1
results
1
Sigma Aldrich N-(3-Formyl-pyridin-4-yl)-2,2-dimethyl-propionamide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.