accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CSiCH3H3COBr

CAS RN 86864-60-0

IUPAC Name: (2-bromoethoxy)(tert-butyl)dimethylsilane
Synonyms: (2-bromoethoxy)(tert-butyl)dimethylsilane
Molecular Weight (g/mol): 239.23
Molecular Formula: C8H19BrOSi
InChi Key: JBKINHFZTVLNEM-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OCCBr
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (4)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)