CAS RN 868696-42-8
IUPAC Name:
5-cyclopropyl-1,2,4-oxadiazol-3-amine
Synonyms:
5-cyclopropyl-1,2,4-oxadiazol-3-amine
Molecular Weight (g/mol):
125.13
Molecular Formula:
C5H7N3O
InChi Key:
OHSZRALIDGBNES-UHFFFAOYSA-N
SMILES:
NC1=NOC(=N1)C1CC1
1
–
1
of
1
results
1
Promotions Available
5-Cyclopropyl-1,2,4-oxadiazol-3-amine, 97%, Thermo Scientific™
5-Cyclopropyl-1,2,4-oxadiazol-3-amine, 97%, Thermo Scientific™
CAS: 868696-42-8 Molecular Formula: C5H7N3O Molecular Weight (g/mol): 125.13 InChI Key: OHSZRALIDGBNES-UHFFFAOYSA-N Synonym: 1,2,4-oxadiazol-3-amine, 5-cyclopropyl PubChem CID: 51072255 IUPAC Name: 5-cyclopropyl-1,2,4-oxadiazol-3-amine SMILES: C1CC1C2=NC(=NO2)N