accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 868755-47-9

IUPAC Name: 2-bromo-1-[4-(furan-2-yl)phenyl]ethan-1-one
Synonyms: 2-bromo-1-[4-(furan-2-yl)phenyl]ethanone
Molecular Weight (g/mol): 265.11
Molecular Formula: C12H9BrO2
InChi Key: SONOLDBWXMKWPY-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C(C=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results