accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 868755-50-4

IUPAC Name: 6-(piperidin-1-yl)pyridine-2-carboxylic acid
Synonyms: 6-(piperidin-1-yl)pyridine-2-carboxylic acid
Molecular Weight (g/mol): 206.25
Molecular Formula: C11H14N2O2
InChi Key: ZBNWTBPSTOBOGV-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC(=CC=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results