accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 868755-51-5

IUPAC Name: 1-[6-(piperidin-1-yl)pyridin-2-yl]methanamine
Synonyms: 1-[6-(piperidin-1-yl)pyridin-2-yl]methanamine
Molecular Weight (g/mol): 191.28
Molecular Formula: C11H17N3
InChi Key: AMSLKMQNMJEDRH-UHFFFAOYSA-N
SMILES: NCC1=NC(=CC=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results