accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNH2

CAS RN 868755-52-6

IUPAC Name: 1-[6-(morpholin-4-yl)pyridin-2-yl]methanamine
Synonyms: 1-[6-(morpholin-4-yl)pyridin-2-yl]methanamine
Molecular Weight (g/mol): 193.25
Molecular Formula: C10H15N3O
InChi Key: JRKWUCQBVPZBBP-UHFFFAOYSA-N
SMILES: NCC1=NC(=CC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results