accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 868755-55-9

IUPAC Name: [3-(1H-1,2,4-triazol-1-yl)phenyl]methanol
Synonyms: [3-(1,2,4-triazol-1-yl)phenyl]methanol
Molecular Weight (g/mol): 175.19
Molecular Formula: C9H9N3O
InChi Key: KUSZGGXHHOYFHS-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC=C1)N1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results