CAS RN 868755-60-6
IUPAC Name:
ethyl 1-methyl-5-(thiophen-2-yl)-1H-pyrazole-3-carboxylate
Synonyms:
ethyl 1-methyl-5-(thiophen-2-yl)pyrazole-3-carboxylate
Molecular Weight (g/mol):
236.29
Molecular Formula:
C11H12N2O2S
InChi Key:
KIYMTNJEKXVPSQ-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=NN(C)C(=C1)C1=CC=CS1
1
–
1
of
1
results
1
Ethyl 1-methyl-5-thien-2-yl-1H-pyrazole-3-carboxylate, 97%, Thermo Scientific™
CAS: 868755-60-6 Molecular Formula: C11H12N2O2S Molecular Weight (g/mol): 236.29 MDL Number: MFCD08060536 InChI Key: KIYMTNJEKXVPSQ-UHFFFAOYSA-N Synonym: ethyl 1-methyl-5-thien-2-yl-1h-pyrazole-3-carboxylate,ethyl 1-methyl-5-thiophen-2-yl pyrazole-3-carboxylate,ethyl 1-methyl-5-2-thienyl pyrazole-3-carboxylate,ethyl 1-methyl-5-thiophen-2-yl-1h-pyrazole-3-carboxylate,1h-pyrazole-3-carboxylicacid,1-methyl-5-2-thienyl-,ethyl ester,1h-pyrazole-3-carboxylicacid, 1-methyl-5-2-thienyl-, ethyl ester PubChem CID: 7537650 IUPAC Name: ethyl 1-methyl-5-thiophen-2-ylpyrazole-3-carboxylate SMILES: CCOC(=O)C1=NN(C)C(=C1)C1=CC=CS1