accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 868755-62-8

IUPAC Name: 5-(furan-2-yl)thiophene-2-carboxylic acid
Synonyms: 5-(furan-2-yl)thiophene-2-carboxylic acid
Molecular Weight (g/mol): 194.2
Molecular Formula: C9H6O3S
InChi Key: ZASUJVQPFQRFPQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(S1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results