accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 868850-50-4

IUPAC Name: 2,6-bis(diphenylamino)-9,10-dihydroanthracene-9,10-dione
Synonyms: 2,6-bis(diphenylamino)anthracene-9,10-dione
Molecular Weight (g/mol): 542.64
Molecular Formula: C38H26N2O2
InChi Key: TZENEWLXCXPNFX-UHFFFAOYSA-N
SMILES: O=C1C2=C(C=C(C=C2)N(C2=CC=CC=C2)C2=CC=CC=C2)C(=O)C2=C1C=C(C=C2)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results