accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 869-10-3

IUPAC Name: 1,6-diethyl 2,5-dibromohexanedioate
Synonyms: 1,6-diethyl 2,5-dibromohexanedioate
Molecular Weight (g/mol): 360.04
Molecular Formula: C10H16Br2O4
InChi Key: UBCNJHBDCUBIPB-UHFFFAOYNA-N
SMILES: CCOC(=O)C(Br)CCC(Br)C(=O)OCC
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)