accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONNNOHOHOOOHONHOOOHO

CAS RN 869-52-3

IUPAC Name: 3,6,9,12-tetrakis(carboxymethyl)-3,6,9,12-tetraazatetradecanedioic acid
Synonyms: 3,6,9,12-tetrakis(carboxymethyl)-3,6,9,12-tetraazatetradecanedioic acid
Molecular Weight (g/mol): 494.45
Molecular Formula: C18H30N4O12
InChi Key: RAEOEMDZDMCHJA-UHFFFAOYSA-N
SMILES: OC(=O)CN(CCN(CCN(CC(O)=O)CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)