accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNSO

CAS RN 869901-02-0

IUPAC Name: tert-butyl 4-(4-formyl-1,3-thiazol-2-yl)piperidine-1-carboxylate
Synonyms: tert-butyl 4-(4-formyl-1,3-thiazol-2-yl)piperidine-1-carboxylate
Molecular Weight (g/mol): 296.39
Molecular Formula: C14H20N2O3S
InChi Key: XLGKMJFDRZHAEV-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)C1=NC(C=O)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results